Learn More
Fisher Science Education™ Digital Tablet Compound Microscope
All-in-one digital microscope is fully functional without computer.
Supplier: Fisher Science Education™ BTW1214RLED
Description
- Integrated tablet for simultaneous viewing through traditional eyepiece
- Pre-loaded Motic Apps for teachers, students and researchers to connect, view and share live images
- Built-in camera
- Wifi interface
Specifications
Coarse and Fine | |
LCD | |
LED | |
Compound Microscope | |
Compound microscopy, view live images on integrated tablet screen, capture images | |
Limited lifetime warranty, excluding bulbs on microscope, 1 year limited warranty on tablet |
1.25 NA Abbe adjustable | |
Monocular | |
4X, 10X, 40XR, 100XR | |
USB: 1280 x720/1280x 1024 preview mode, 2592x1944 (5MP) still image; Wifi: 1280 x960 preview mode, 1280 x960 (1.3MP) still image | |
Reversed quadruple revolving |