Learn More
Fisher Scientific™ ChemExpress™ Electronic MSDS System
MSDS information at your fingertips!
 View more versions of this product
Catalog No. 07-703-148
This product is either new or inactive, and cannot be ordered online.
Description
Description
CD-ROM provides critial safety information on over 3000 analytical laboratory reagents. All MSDSs are in the internationally recognized 16-section ANSI standard format, so finding MSDSs is quick and easy. An updated CD-ROM is sent every three months.
  • Features searches by full or partial chemical name, catalog number, molecular formula, and CAS number of pure product or component
  • System includes six additional languages (French, Spanish, German, Italian, Dutch, and Swedish), and also allows additional MSDSs to be added
  • Compatible with all versions of Windows™ (Minimum system requirements for operation: Pentium™ processor, 32MB RAM, CD-ROM drive and mouse, hard drive with 100-600MB free space, and monitor with VGA or higher resolution)
  • May be installed to hard drive for faster performance, or run from the CD
  • Fisher/Acros™ Organics/Maybridge Version includes same features as Fisher Chemical edition, but also provides access to Acros Organics and Maybridge MSDSs for 26,000 organic and analytical laboratory reagents
  • Both editions available configured for single user, or for network that can be accessed by an unlimited number of users
Specifications
Specifications
CD-ROM
Electronic MSDS System
Provides access to MSDS for Fisher Chemicals/ACROS Organics/Maybridge
Single user
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.