Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Scientific™ Chemical-Resistant Check Valves

Constructed of chemical-resistant Kynar™ body and Viton™ valve

 View more versions of this product

Catalog No. 15-078-209


  Check Availability
Add to cart

Specifications

Description

Used with acids, bases, water, chemical processing and alcohols. Prevents damage to equipment, pumps, and flow systems by preventing backflow.

  • Clear flow path
  • Inert high purity smooth
  • Operating ranges: 0 to 50psi and -40° to +126°C
  • Autoclavable; zero maintenance
  • 1.25 in. length unit installs in any position

Description & Specifications

Specifications

Check Valve
4.74mm
Yes
0.187 in.
Kynar™ and Viton™
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit