Learn More
Fisherbrand™ Circular Bottom Desi-Vac™ Container Desiccators with Vacuum Pump
Desiccator with built-in manual vacuum pump eliminates need for separate pump, tubing and stopcock
 View more versions of this product
Catalog No. 08-664-5B
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
Do not autoclave or expose container to organic solvents or their vapors. Do not use concentrated acids as desiccants.
Description
Description
Desiccator with built-in manual vacuum pump eliminates need for separate pump, tubing and stopcock
  • Ideal for drying, preserving, storing and shipping samples
  • Self-contained unit holds a vacuum of 14.7 in.Hg (373mm) in excess of 24 hours
  • Rubber O-ring in cover ensures a tight seal
  • To remove cover, simply push release button to allow air into container
  • Clear polycarbonate body, metal pump handle, chemical-resistant ABS plastic pump and cover
  • No plate required
Ideal for drying, preserving, storing and shipping samples
Specifications
Specifications
28.74 oz.
Desiccator
Metal pump handle, chemical-resistant ABS plastic vacuum pump and cover, release button on top, O-Ring-sealed cover
4.12 in.
5 in.
NA
850mL
Circular
Polycarbonate
10.4cm
12.7cm