Learn More
Fisher Scientific™ CMU Metal Controlled and Uncontrolled Heating Mantles
Features aluminum housing that is tough, chemical-resistant, lightweight and easy to clean
 View more versions of this product
Catalog No. 11-747-200
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
1.6 oz., 50mL
Heating Mantle
842°F
75w
0.75kg
17.5cm
50mL
CE, CSA
115V 50/60Hz, 75w
450°C
26.45 oz.
6.88 in.
1.6 oz.
115V