Learn More
Fisher Scientific™ CMU Metal Controlled and Uncontrolled Heating Mantles
Features aluminum housing that is tough, chemical-resistant, lightweight and easy to clean
 View more versions of this product
Catalog No. 11-474-63
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
17 oz., 500mL
Heating Mantle
842°F
280w
1.5kg
22cm
500mL
CE, CSA
230V 50/60Hz, 220w
450°C
52.91 oz.
8.66 in.
17 oz.
230V
Accessories