Fisher Scientific™ DAS-3 Data Acquisition System for Fisher Scientific™ Traceable™ Products with Computer Output
Powerful and easy to use computer data capture program works with Traceable Instruments with computer output—designed to work with Windows™ 98/Me/NT/2000/XP/Vista/Windows 7. Connects up to eight instruments all running simultaneously on one computer.
 View more versions of this product
Catalog No. 15-077-73
  Check Availability
Add to Cart
Includes: CD, 6 ft. cable (supplied USB, serial, and instrument connections) cable plugs into the instrument and computer.
    Specifications
    Product Type Computer data acquisition system
    For Use With (Application) Capture readings permanently, displays minimum/maximum readings
    For Use With (Equipment) Manometer/pressure/vacuum gauges, memory hygrometer/thermometer, expanded range conductivity meter and memory hygrometer/thermometer
    Includes CD, 6 ft. USB cable
    Type Computer data acquisition system
    • Capture readings permanently
    • Records in intervals from 1 to 10,000 seconds
    • Straightforward plug-and-play
    • Displays minimum/maximum readings
    • Utilizes an alarm mode that permits the user to be notified visually, audibly, and by email when an alarm is triggered
    • Unlimited readings are saved to a file that can be viewed and printed as is, or may be printed in any report or spreadsheet format
    • Networking server/client capability allows the captured data to be monitored on a remote workstation
    • Readings may be streamed to cell phones or anywhere via email every minute or in any time interval
    • 24/7 notification of audible/visual alarms are sent to user's computer, remote computer, via email anywhere, and by email to cell phones
    • Program automatically installs in less than two minutes—no user programming, no user keystrokes, no user entries required: simply connect the cable to the instrument and to a USB port (or serial port) and data is captured
  • Description & Specifications
Related Products
Product Content Feedback