Learn More
Fisher Science Education™ Series Digital Compound Microscope Products for Science Education
Designed for advanced high school, university and laboratory use. Fisherbrand™ DC20-169 Series Digital Compound Microscope, an advanced compound microscope with a built-in HDMI 1080 camera.
Manufacturer:  Fisher Science Education™ DC20169SP
 View more versions of this product
Catalog No. S24751
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The DC20-169 Digital compound microscope is a perfect for teachers who want to project high-quality live images directly to a monitor or LCD projector.
Microscope features:
  • Binocular viewing head with 30 degree inclined eyepieces for less eye strain.
  • Energy-efficient LED illumination
  • New rackless stage for ease of operation
  • Upgraded objectives for image clarity with your choice of ASC, Semi-plan, plan or plan phase options
Camera specifications:
  • Digital interface:  USB 2.0 and HDMI
  • Max resolution:  up to 2.0MP (captured to SD card).   Live USB output 2.0MP
  • Operating Requirements:  Windows 7 or higher, OSX and 250 MB of free HD space
Specifications
Specifications
Education
DC20-169- SP
1.25 Abbe N.A.
Semi-plan
LED
Microscope
Focus control: Coaxial
Built-in HDMI
40X, 100X,400X, 1000X
Binocular
Limited lifetime on the microscope, 1 year limited warranty on camera