Fisher Scientific™ Digital Compound Microscopes
Available with 3.0MP or 1080 via HDMI
 View more versions of this product
Catalog No. 11-350-139
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Condenser Type Focusable Abbe Condenser N.A. 1.25
    Objective Types Plan 4X, 10X, 40X, 100X
    Eyepiece Widefield 10X/18mm
    For Use With (Application) University and laboratory use
    Lighting Type 12V 20w halogen w/variable rheostat control
    Nosepiece Reversed quadruple revolving nosepiece
    Weight (Metric) 6.3kg
    Power Supply Details IEC 110/220V (UL)
    Warranty Microscope: 5 year limited warranty, excluding bulbs, cords, and fuses; Camera: 1 year
    Product Type Digital compound microscope with 5.0MP digital camera
    Height (Metric) 405mm
    Includes IEC Power Cord, Dustcover, Blue, Green, and Yellow Filters
  • Description & Specifications
Related Products
Product Content Feedback