Learn More
Fisher Scientific™ Digital Compound Microscopes
Available with 3.0MP or 1080 via HDMI
 View more versions of this product
Catalog No. 11-350-140
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
405mm
University and laboratory use
Plan phase 4X, 10X, 40X, 100X
12V 20w halogen w/variable rheostat control
6.3kg
Microscope: 5 year limited warranty, excluding bulbs, cords, and fuses; Camera: 1 year
IEC Power Cord, Dustcover, Blue, Green, and Yellow Filters
Focusable Abbe Condenser N.A. 1.25
Widefield 10X/18mm
Reversed quadruple revolving nosepiece
IEC 110/220V (UL)
Digital compound microscope with 5.0MP digital camera