Fisher Scientific™ Disposable Generators For Homogenizer Model 125
For processing of multiple samples per day (more than 50)
 View more versions of this product
Catalog No. 08-451-678
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Diameter (English) Shaft 3.3 in.
    Diameter (Metric) Shaft 8.4cm
    For Use With (Application) Ideal for dispersing plant, animal or human tissue in small amount of liquid
    Packaging Quantity 25/Pk.
    Capacity (Metric) 0.3 to 10mL
    For Use With (Equipment) Model 125 homogenizer
    Type Disposable Generator
  • Description & Specifications
Related Products
Product Content Feedback