Learn More
Fisherbrand™ Dual-head Vacuum Pumps
Ideal for portable applications
$780.50
Specifications
Maximum Pressure 18 psi
View More Specs
Products 1
Catalog Number Air Displacement Electrical Requirements Horsepower Motor Type Price Quantity & Availability  
Catalog Number Air Displacement Electrical Requirements Horsepower Motor Type Price Quantity & Availability  
13-310-904
View Documents
22.6 L/min. 230 V 50 Hz 0.9 A 1/45 hp TEFC
Each for $780.50
Add to cart
Description
Description
  • Increased capacity over single-head pump
  • Use two pump heads independently or connect them in series with 3/8 in. ID Tygon™ vacuum tubing for up to 20 in. Hg vacuum
  • Includes: PTFE thread tape, four PE 3/8 in. NPT(M) x 3/8 in. hose barb adapters, and 6 ft. three-wire cord with plug
Specifications
Specifications
18 psi