Learn More
Fisher Scientific™ Ever-Safe™ Thermometers Products for Science Education
Safer for students and the environment.
Includes:
  • No-roll fitting
  • Storage tube
  • Certificate of Accuracy
Products 1
Catalog Number Description Price Qty Availability  
Catalog Number Description Price Qty Availability  
S63351 Products for Science Education
View Documents
N/A Total immersion; Range: 0° to +230°F This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Safer than toxic mercury and even spirit-filled thermometers
  • Filled with a biodegradable liquid and dye
  • Easy-to-read blue liquid filling is completely nontoxic