Learn More
Fisherbrand™ Exhaust Oil Recycler with Built-in Filter Element
Economical mist eliminator is recommended for use when operating the pump at a continuous vacuum pressure of 1 torr to 10 torr
$722.00
Specifications
Includes | Adapters |
---|---|
Type | Exhaust Oil Recycler with Built-in Filter Element |
Catalog Number | Diameter (English) Outside | Diameter (Metric) Outside | Overall Height (English) | Overall Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Diameter (English) Outside | Diameter (Metric) Outside | Overall Height (English) | Overall Height (Metric) | Price | Quantity & Availability | |||
01-257-7A
|
2-3/4 in. | 70 mm | 6.25 in. | 15.9 cm |
Each of 1 for $722.00
|
|
|||
Description
Continuously separates air and oil from the pump exhaust, returning captured oil to the pump via the gas ballast inlet (when gas ballast valve is open). This process effectively reduces oil consumption and minimizes maintenance requirements by extending the time between oil changes.
Specifications
Adapters | |
Exhaust Oil Recycler with Built-in Filter Element |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.