Learn More
Fisherbrand™ Filter/Regulator and Lubricator Complete Assembly for Air Stirrers
For long and trouble-free life of air-powered stirrers.
Catalog No. 14-512-900
Add to cart
Description
Description
Filter removes oil and water aerosols and foreign particles larger than 40 microns in the air line
  • Integral filter/regulator and lubricator combination
  • With 1.5 in. diameter gauge, 0-160 psi
Order Info
  • Automatic air line lubricator must be installed in the air line just ahead of the motor
  • Lubricator should be adjusted to feed one drop of oil for every 50-70 cfm of air groung through the motor (approximately 1 drop every five minutes)
Specifications
Specifications
Lubricator/Filter/Regulator Assembly
Long and trouble-free life of air-powered stirrer
Air Stirrer
1.5 in. dia. Gauge