Learn More
Fisherbrand™ Flask Clamps for Incubating Mini Shakers
Available in stainless steel and PVC formats.
Description
- PVC clamps are autoclavable and can be used with polycarbonate flasks
- Available in range of volumes from 10mL to 250mL
Specifications
25 mL Flask | |
Clamp | |
Stainless Steel | |
Incubating mini shaker |