Learn More
Fisher Scientific™ General-Purpose Undercounter Freezer
Convenient storage in small spaces
Catalog No. 97-926-1
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Manual defrost
  • R-134a CFC-free refrigerant
  • Hydraulic thermostat
  • Baked-on, acrylic enamel exterior
  • Easy-to-clean formed interior
  • Solid door is mounted to open from left
Warranty and Services
Two years parts, one year labor
Documents
Accessories