Learn More
Fisher Scientific™ Generators For Homogenizer Model 1000
Available with high performance sleeve bearings made from Valflon™
 View more versions of this product
Catalog No. 08-451-682
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
7.87 in.
Ideal for dispersing and mixing of various components to create finest homogenates, emulsions, suspensions, accelete reactions
Dim (Rotor/Stator/Shaft): 23/30/200mm
Model 1000 homogenizer
20cm
Stainless Steel
300 to 7000mL
Generator