Fisher Scientific™ Handheld Refractometers
Nine different handheld models
 View more versions of this product
Catalog No. 13-946-25
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Includes Soft-sided, Foam lined storage case with instruction manual and calibration screwdriver
    Increments 0.1°
    Temperature Compensation 10 and 30°C
    Accuracy ±0.1°
    Scale Range 0 to 10° Brix with ATC
  • Description & Specifications
Related Products
Product Content Feedback