Fisher Scientific™ Hands-Free Fold-Up Magnifiers
Allow two-handed examination of objects while yielding a sharp, crisp and distinct image
 View more versions of this product
Catalog No. 14-648-21
  Check Availability
Add to Cart
    Specifications
    Diameter (English) Lens 3 in.
    Diameter (Metric) Lens 7.62cm
    For Use With (Application) For use in quality control, labs, food, electronics, manufacturing and universities
    Material Glass
    Magnification 3x
    Product Type Hands-Free™ Fold-up Magnifier
    Fisher Scientific™ Hands-Free Fold-Up Magnifiers all fold twice for compact storage. Allow two-handed examination of objects while yielding a sharp, crisp and distinct image.
    • Bottom panel features white grid markings in both inches and millimeters to assist in measuring
    • All-glass optics ensure perfect focus with zero distortion
    • Seven sizes are available, including three featuring brilliant battery-powered illumination
  • Description & Specifications
Related Products
Product Content Feedback