Learn More
Fisher Scientific™ Heating and Stirring Mantles
Chemically resistant polypropylene outer casing. Electrothermal™ Heating and Stirring Mantles offer single or bi-directional stirring.
 View more versions of this product
Catalog No. 11-473-19
Due to product restrictions, please to purchase or view availability for this product.
Specifications Description & Specifications
Specifications
3.3 oz., 100mL
96w
1.73kg
17.5cm
100mL
CE
61.02 oz.
6.88 in.
3.3 oz.
115V
Accessories