Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ High Form Vinyl Chair with Casters, Medium Bench Height Available on GSA/VA Contract for Federal Government customers only.

Choose to outfit your workplace with Fisherbrand chairs - biomechanically engineered seating offered in many configurations.

Catalog No. 14-360-119


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Burgundy
24 to 31 in.
60.9cm
Steel Round Tube
12 in.
21 to 28 in.
21 to 28 in.
35.5 to 45.7cm
45.7cm
High-Form
Chair
17 in.
19 in.
48.2cm
48.2cm
Fixed
Standard Lift
Round tube
24 in.
Casters
Powder-Coated Steel
14 to 18 in.
30.4cm
53.3 to 71.1cm
18 in.
Pneumatic
Medium Bench Height
Vinyl
19 in.
43.1cm
Yes
Yes
Chair
Vinyl Upholstered Foam
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit