Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Fisher Scientific™ HistoPrep™ Stainless-Steel Base Molds
Smooth finish and rounded corners make cleanup fast and easy
 View more versions of this product
Catalog No. 15-182-505E
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
1.45 x 0.94 x 0.19 in. (3.7 x 2.4 x 0.5cm)
0.19 in.
Stainless Steel
1.45 in.
0.94 in.
12 Pack
Designed for use with commonly used embedding equipment for superb thermal exchange and durability
0.5cm
Base Mold
3.7cm
2.4cm
Accessories