Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bio Plas™ HistoPlas™ Tissue Processing Capsules
The integral lid fits tightly—you never need to worry about losing your tissues
$906.00
Catalog Number | Mfr. No. | Diameter (English) | Depth (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (English) | Depth (English) | Price | Quantity & Availability | ||||
15-182-218
|
Bio Plas™
6010 |
1.5 in. | 0.33 in. |
Case of 1000 for $906.00
|
|
15-182-219
|
Bio Plas™
6020 |
1 in. | 0.25 in. |
Case of 1000 for $906.00
|
|
Description
The integral lid fits tightly—you never need to worry about losing your tissues
- Disposable polypropylene capsules ensure proper fluid exchange in processing and infiltration
- Open-mesh base and lid allow superior flow and drainage
- One-piece design is hinged for perfect lid/base alignment
- With generous labeling surface for quick identification
Specifications
Tissue Capsule | |
White | |
1000/Cs. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title