Fisher Scientific™ Homogenizer Model 125 Package
For fast and economical dispersion of both small and large amount of products
 View more versions of this product
Catalog No. 08-451-662
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Type Homogenizer Package
    Voltage 115V
    For Use With (Application) Ideal for production of dispersions and emulsions
    Includes Model 125 homogenizer (115V), stand, 5,7 and 10mm stainless steel generators, one box of disposable 7mm generator
  • Description & Specifications
Related Products
Product Content Feedback