Learn More
Fisher Scientific™ Horizon™ Performance Series Model 853VES Plasmafuge-12
Medium capacity 12-place horizontal centrifuge with digital controls
 View more versions of this product
Catalog No. 22-029-422
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
5999rpm
No
115/230V
23cm
39cm
Yes, Electric; 1 to 99 min.
32cm
Electronic
13.7kg
115/230V 50/60Hz
5,000 to 5,999rpm
3000 to 3999 x G
50/60Hz
8.5 in.
15.5 in.
Up to 10 memory settings
12.5 in.
Yes
30 lb.
15.5 x 12.5 x 8.5 in. (39 x 32 x 23cm)
3,000 to 3,999 x G
Yes
Accessories