Learn More
Fisher Scientific™ Incubating/Cooling Mini Shakers
Microplate ready without needing additional accessories
 View more versions of this product
Catalog No. 02-217-756
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications Description & Specifications
Specifications
1A
41 to 104°F
200w
1 sec. to 160 Hours
10.5 in.
Polycarbonate lid
45.5cm
Two microplates/modular blocks
27.9cm
80%
5 to 40°C
230V
LEDLED
26.7cm
17.9 in.
100 to 1200rpm
11 in.
17.9 x 11 x 10.5 in. (45.5 x 27.9 x 26.7cm)