Learn More
Fisher Scientific™ Isotemp™ 4100 H12A Open Heated Acrylic Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Acrylic open heated baths are fitted with bridge plates for placement of the control head
 View more versions of this product
Catalog No. 13-874-178
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
10 to 80°C
8 to 12L
60Hz
33.7cm
34.7cm
35.3cm
13.7 x 13.9 x 13.3 in. (34.7 x 35.3 x 33.7cm)
4100 H12A
310mbar
7.6kg
16.8 lb.
Isotemp™ open heated Acrylic bath circulator
270.5 to 405.7 oz.
115V
13.3 in.
13.7 in.
13.9 in.
115V 60Hz
1
17L/min.
Force
1000w
±0.1°C