Learn More
Fisher Scientific™ Isotemp™ 4100 H19A Open Heated Acrylic Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Acrylic open heated baths are fitted with bridge plates for placement of the control head
 View more versions of this product
Catalog No. 13-874-179
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
10 to 80°C
12 to 19L
60Hz
33.7cm
52.5cm
35.3cm
20.7 x 13.9 x 13.3 in. (52.5 x 35.3 x 33.7cm)
4100 H12A
310mbar
8.75kg
19.3 lb.
Isotemp™ open heated Acrylic bath circulator
405.7 to 642.4 oz.
115V
13.3 in.
20.7 in.
13.9 in.
115V 60Hz
1
17/min.
Force
1000w
±0.1°C