Learn More
Fisher Scientific™ Isotemp™ 4100 H6A Open Heated Acrylic Bath Circulators Available on GSA/VA Contract for Federal Government customers only.
Acrylic open heated baths are fitted with bridge plates for placement of the control head
 View more versions of this product
Catalog No. 13-874-047
  Check Availability
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Specifications Description & Specifications
Specifications
10 to 80°C
5.5 to 7.0L
50Hz
33.7cm
40.6cm
18.7cm
16 x 7.4 x 13.3 in. (40.6 x 18.7 x 33.7cm)
17L/min.
Force
13 lb.
Isotemp™ open heated Acrylic bath circulator
185.9 to 236.6 oz.
230V
13.3 in.
16 in.
7.4 in.
230V 50Hz
1.2
310mbar
5.8kg
±0.5°C