Fisher Scientific™ Isotemp™ Analog Dry Bath Incubators, International
Precise temperature control resolution and a much smaller footprint than a general purpose incubator
 View more versions of this product
Catalog No. 17-151-450
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Warranty 2 Years
    Wattage 100w
    Temperature Range (Metric) 5° to 130°C
    Certifications/Compliance CE, CSA
    Display LED
    Includes 1 Block Accommodation
    Temperature Uniformity ±0.5 at 37°C
    Voltage 240VAC
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback