Learn More
Fisher Scientific™ Isotemp™ Basic Magnetic Stirrers
Compact, low-profile stirrers with analog control
 View more versions of this product
Catalog No. 11-100-49S
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
60 to 1200rpm
Ceramic
120V 50/60Hz
1
7.25 x 7.25 in. (18.4 x 18.4cm)
Accessories