Fisher Scientific™ Isotemp™ Basic Magnetic Stirrers
Compact, low-profile stirrers with analog control
 View more versions of this product
Catalog No. 11-102-16S
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Stirring Positions 1.0
    Top Plate Surface Area (English) 4.25 x 4.25 in. (10.8 x 10.8cm)
    Electrical Requirements 230V 50/60Hz
    Speed 60 to 1200rpm
    Voltage 230V
    Material Ceramic
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback