Learn More
Fisher Scientific™ Isotemp™ Basic Magnetic Stirrers
Compact, low-profile stirrers with analog control
 View more versions of this product
Catalog No. 11-102-49S
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
60 to 1200rpm
11kg
Ceramic
230V 50/60Hz
1.0
230V
7.25 x 7.25 in. (18.4 x 18.4cm)
Accessories