Learn More
Thermo Scientific™ Thermo Scientifi™ Chiller External Pressure Reducers
External pressure reducers
Supplier: Thermo Scientific™ 196000000001
Description
External pressure reducers for ThermoChill Recirculating chillers
- 1/2 in. external pressure reducer
- Adjustable to safely reduce or limit system pressure and flow
Specifications
External Pressure Reducer for ThermoChill III | |
ThermoChill III Recirculatig chiller | |
External Pressure Reducer (EPR) | |
1/2 in. |