Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Isotemp™ Digital and Analog Dry Bath Incubators

Provide controlled dry heat for a wide variety of clinical and general chemistry applications

 View more versions of this product

Catalog No. 11-718Q

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: Digital Models come with temperature probe, line cord and plug. Analog Models include high and low thermostat, power switch, pilot light, line cord and plug.

Requires : Heating blocks, sold separately, hold regular tubes, microcentrifuge tubes, square cuvets, or microplates.

Description

Description

Choice of 1- to 6-block Analog Models, 2- or 4-block Digital Models.

  • Exceptional temperature control and uniformity
  • Temperature range of 5°C above ambient to 130°C
  • Powder-coated steel body construction provides durability for demanding applications
  • Compact design saves valuable bench space
  • Choice of interchangeable heating blocks

Digital Dry Baths with Microprocessor Controller

  • PID microprocessor control, digital temperature setting
  • Just press up/down arrows to set temperature in 0.1° increments
  • Effectively hold samples at constant, warm temperature
  • 5°C above ambient to 130°C range; ±0.4° uniformity at 37°C; stability, ±0.2°C
  • Bright LED display continuously shows actual temperature in °C
  • Temperature probe in bottom of bath provides improved accuracy and control; free probe included
  • Model 125DQ holds two heating blocks; Model 145DQ holds four heating blocks in 2 x 2 configuration
Analog Dry Baths with Dual-thermostat Controls
  • Sloped control panel makes controls highly visible
  • Dual bimetallic thermostat controls
  • Stability is ±1.5°C, uniformity ±0.5°C at 37°C
  • Use thermometer to read current temperature of blocks, then adjust control to reach desired setpoint
  • Efficient design: operate at substantially lower external temperature than other models

Applications:

Ideal for enzyme reactions, inactivation of sera, Rh studies, blood crossmatching and cholesterol determinations

†International model

Order Info

All models accept 2.91L x 3.75W x 2 in.H (7.4 x 9.5 x 5.1cm) blocks, sold separately.

Certifications

Digital Models are CSA approved. Analog Models are UL listed.

Warranty and Services

One year

Specifications

Specifications

5 to 130°C
120V
8.6cm
19.3cm
15.7cm
50/60Hz
UL
120V 50/60Hz
LED
5 lb.
7.6 x 6.2 x 3.4 in. (32.8 x 28.6 x 8.6cm)
32.8cm
28.6cm
8.6cm
±0.5 at 37°C
3.38 in.
7.63 in.
6.38 in.
100w
0.83A
Powder coated Steel
7.63 x 6.38 x 3.38 in. (19.3 x 15.7 x 8.6cm)
Dual bimetallic thermostat controls (high and low), power switch, pilot light, line cord and plug
2kg
7.6 in.
6.2 in.
3.4 in.
1 Year
Documents
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit