Learn More
Fisher Scientific™ Isotemp™ Digital-Control Water Baths: Model 202
Ideal for industrial, clinical, pharmaceutical or biomedical procedures
 View more versions of this product
Catalog No. 15-462-2Q
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Ambient to 100°C
7.62 in.
50/60Hz
25cm
7.25 lb.
10.75 in.
1.7A
CSA
5.25 x 5.88 x 6 in. (13 x 14.8 x 15cm)
±0.5°C
Acrylic cover
±0.5°C
120V
19cm
9.88 in.
202
3.3kg
27cm
2L
9.88 x 10.75 x 7.62 in. (25 x 27 x 19cm)
Microprocessor PID
Industrial, clinical, pharmaceutical or biomedical procedures
0.1°C/F
1 Year
Accessories