Learn More
Fisher Scientific™ Isotemp™ Digital-Control Water Baths: Model 202S
Ideal for industrial, clinical, pharmaceutical or biomedical procedures
 View more versions of this product
Catalog No. 15-462-3SQ
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Ambient to 100°C
6.25 in.
60Hz
27cm
8.88 lb.
14.17 in.
1.7A
CSA
Microprocessor PID
Industrial, clinical, pharmaceutical or biomedical procedures
±0.1°C
1 Year
230V
16cm
10.5 in.
202s
4kg
36cm
2L
10.5 x 14.17 x 6.25 in. (27 x 36 x 16cm)
±0.5°C
Acrylic cover
±0.5°C
Accessories