Learn More
Fisher Scientific™ Isotemp™ Digital Magnetic Stirrers
Compact, precise, and perfect for timed applications
 View more versions of this product
Catalog No. 11-300-49S
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
60 to 1200rpm
120V
3.875 in.
12.75 in.
8.5 in.
50/60Hz
18.4cm x 18.4cm
18.4cm
18.4cm
12.75 x 8.5 x 3.875 in. (27.9 x 18.7 x 8.4cm)
11kg
Ceramic
8.4cm
27.9cm
18.7cm
7.25 in. x 7.25 in.
7.25 in. x 7.25 in.
7.25 in.
120V 50/60Hz
Digital Magnetic Stirrer
Accessories