Learn More
Fisher Scientific™ Isotemp™ Direct Heat CO2 Incubators
Discover exceptional air purity with in-chamber HEPA air filtration
 View more versions of this product
Catalog No. 20-235-33
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Stainless Steel
39.5 in.
25 in.
120V
6.5 cu. ft.
6.5 cu. ft., 184L
IR sensor
95%
66.8cm
Ambient 5 to 60°C
100.3cm
63.5cm
50/60Hz
184L
120V 50/60Hz
0 to 20%
26.3 in.
1 Year