Learn More
Fisher Scientific™ Isotemp™ Economy Analog-Control Water Baths
Economical, general-purpose water baths are ideal for industrial, clinical, pharmaceutical and biomedical procedures
 View more versions of this product
Catalog No. 15-460-2SQ
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
5 to 70/100°C
6.25 in.
50/60Hz
27cm
7.25 lb.
14.17 in.
2L
10.5 x 14.17 x 6.25 in. (27 x 36 x 16cm)
Industrial, clinical, pharmaceutical or biomedical procedures
±0.2°C
120V
16cm
10.5 in.
102S
3.3kg
36cm
CSA
Analog
Sample tray, thermometer clip, nonhazardous liquid, partial immersion (76mm) thermometer (-10 to 110°C), line cord and grounded NEMA 5-15P plug
1 Year
Accessories