Fisher Scientific™ Isotemp™ Economy Analog-Control Water Baths

Economical, general-purpose water baths are ideal for industrial, clinical, pharmaceutical and biomedical procedures

 View more versions of this product

Catalog No. 15-460-10Q

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Warranty 1 Year
    Length (Metric) Exterior 42cm
    Model 110
    Temperature Range (Metric) 5 to 75/100°C
    Width (English) Exterior 15.38 in.
    Amperage 5.8A
    Capacity (English) 2.64 gal.
    Dimensions (L x W x H) Exterior 16.53 x 15.38 x 8.88 in. (42 x 39 x 23cm)
    Voltage 120V
    Height (English) Exterior 8.88 in.
    Height (Metric) Exterior 23cm
    Hertz 50/60Hz
    Length (English) Exterior 16.53 in.
    Shipping Weight (English) 16.5 lb.
    Shipping Weight (Metric) 7.5kg
    Width (Metric) Exterior 39cm
    Capacity (Metric) 10L
    Certifications/Compliance CSA
    Capacity 2.64 gal., 10L
    Electrical Requirements 120V 50/60Hz
    Controller Type Analog
    For Use With (Application) Industrial, clinical, pharmaceutical or biomedical procedures
    Includes Sample tray, thermometer clip, nonhazardous liquid, partial immersion (76mm) thermometer (-10 to 110°C), line cord and grounded NEMA 5-15P plug
    Temperature Stability ±0.2°C

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit