Fisher Scientific™ Isotemp™ Economy Analog Control Water Baths 230V Models

Easy-to-use, front-mounted controls

 View more versions of this product

Catalog No. 15-460-11Q

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »

Alert

Not for sale in North America.


    Specifications

    Warranty 1 Year
    Temperature Range (Metric) 100°C
    Width (Metric) Exterior 39cm
    Amperage 3A
    Certifications/Compliance CSA
    Capacity 2.64 gal., 10L
    Height (English) Exterior 8.88 in.
    Height (Metric) Exterior 23cm
    Length (English) Exterior 16.53 in.
    Length (Metric) Exterior 42cm
    Model 110
    Shipping Weight (Metric) 7.5kg
    Width (English) Exterior 15.38 in.
    Capacity (English) 2.64 gal.
    Dimensions (L x W x H) Exterior 16.53 x 15.38 x 8.88 in. (42 x 39 x 23cm)
    Electrical Requirements 230V 50/60Hz
    Controller Type Analog
    Voltage 230V
    For Use With (Application) Industrial, clinical, pharmaceutical or biomedical procedures
    Includes Line cord and European style CE plug, sample tray, thermometer clip and nonhazardous-liquid, partial immersion (76mm) thermometer (-10 to 110°C) come with each bath
    Temperature Stability ±0.2°C
    Hertz 50/60Hz
    Shipping Weight (English) 16.5 lb.
    Capacity (Metric) 10L

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit