Fisher Scientific™ Isotemp™ Economy Analog Control Water Baths 230V Models

Easy-to-use, front-mounted controls

 View all options for this product

Catalog No. 15-460-16Q


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Temperature Range (Metric) 100°C
    Voltage 230V
    Height (English) Exterior 8.88 in.
    Height (Metric) Exterior 23cm
    Hertz 50/60Hz
    Length (English) Exterior 16.53 in.
    Length (Metric) Exterior 42cm
    Model 115
    Shipping Weight (English) 24.5 lb.
    Shipping Weight (Metric) 11.1kg
    Width (English) Exterior 24.38 in.
    Width (Metric) Exterior 62cm
    Amperage 4.3A
    Capacity (English) 1.32 and 2.64 gal.
    Capacity (Metric) 5 and 10L
    Certifications/Compliance CSA
    Dimensions (L x W x H) Exterior 16.53 x 24.38 x 8.88 in. (42 x 62 x 23cm)
    Electrical Requirements 230V 50/60Hz
    Controller Type Analog
    For Use With (Application) Industrial, clinical, pharmaceutical or biomedical procedures
    Includes Line cord and European style CE plug, sample tray, thermometer clip and nonhazardous-liquid, partial immersion (76mm) thermometer (-10 to 110°C) come with each bath
    Temperature Stability ±0.2°C
    Warranty 1 Year

    Economical, general-purpose water baths are ideal for industrial, clinical, pharmaceutical, and biomedical procedures. Seven models, including shallow form and dual chamber, with capacities from 2 to 28L. Shallow-form Model 102S is perfect for heating samples in microcentrifuge tubes and for other molecular biology procedures. Model 115 has two chambers, each with its own temperature control panel.

    • Precise temperature control up to 100°C (212°F)
    • Capacities from 2 to 28L
    • Easy-to-use controls on front panel for full access and safe operation
    • Dual thermostats guard against thermal runaway; elements won't burn out even if bath runs dry
    • Hinged see-through acrylic gable covers have unique fins that keep hands away from hot vapors; stay open at 90° position or lift off to accommodate large glassware
    • Corrosion-resistant exterior with acrylic enamel finish; temperature-resistant plastic rim fits snugly to insulate and prevent leaks
    • Outside of the bath stays cool to the touch even after hours of operation
    • Low-profile design makes access convenient
    • Deep-drawn stainless-steel reservoirs are seamless to prevent leaks; rounded corners make cleaning easy

    European-style plug

  • Description & Specifications
  • Accessories

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit