Learn More
Fisher Scientific™ Isotemp™ Economy Analog Control Water Baths 230V Models
Easy-to-use, front-mounted controls
 View more versions of this product
Catalog No. 15-460-3Q
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
Not for sale in North America.
Specifications Description & Specifications
Specifications
100°C
7.48 in.
50/60Hz
25cm
7.25 lb.
10.75 in.
0.8A
CSA
Analog
Line cord and European style CE plug, sample tray, thermometer clip and nonhazardous-liquid, partial immersion (76mm) thermometer (-10 to 110°C) come with each bath
1 Year
230V
19cm
9.88 in.
102
3.3kg
27cm
2L
9.88 x 10.75 x 7.48 in. (25 x 27 x 19cm)
Industrial, clinical, pharmaceutical or biomedical procedures
±0.2°C