Fisher Scientific™ Isotemp™ Economy Analog Control Water Baths 230V Models

Easy-to-use, front-mounted controls

 View more versions of this product

Catalog No. 15-460-3SQ

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »

Alert

Not for sale in North America.


    Specifications

    Warranty 1 Year
    Length (English) Exterior 10.5 in.
    Temperature Range (Metric) 100°C
    Width (Metric) Exterior 36cm
    Amperage 0.8A
    Capacity (Metric) 2L
    Certifications/Compliance CSA
    Capacity 0.52 gal., 2L
    Electrical Requirements 230V 50/60Hz
    Includes Line cord and European style CE plug, sample tray, thermometer clip and nonhazardous-liquid, partial immersion (76mm) thermometer (-10 to 110°C) come with each bath
    Height (Metric) Exterior 19cm
    Length (Metric) Exterior 27cm
    Model 102S
    Width (English) Exterior 14.17 in.
    Capacity (English) 0.52 gal.
    Dimensions (L x W x H) Exterior 10.5 x 14.17 x 7.48 in. (27 x 36 x 19cm)
    Controller Type Analog
    Voltage 230V
    For Use With (Application) Industrial, clinical, pharmaceutical or biomedical procedures
    Temperature Stability ±0.2°C
    Height (English) Exterior 7.48 in.
    Hertz 50/60Hz
    Shipping Weight (English) 8.88 lb.
    Shipping Weight (Metric) 4kg

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit