Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
Fisher Scientific™ Isotemp™ Economy Analog Control Water Baths 230V Models
Easy-to-use, front-mounted controls
 View more versions of this product
Catalog No. 15-460-6Q
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Not for sale in North America.
Specifications Description & Specifications
Specifications
100°C
7.48 in.
50/60Hz
27cm
9.88 lb.
14.17 in.
1.3A
CSA
Analog
Line cord and European style CE plug, sample tray, thermometer clip and nonhazardous-liquid, partial immersion (76mm) thermometer (-10 to 110°C) come with each bath
1 Year
230V
19cm
10.5 in.
105
4.5kg
36cm
5L
10.5 x 14.17 x 7.48 in. (27 x 36 x 19cm)
Industrial, clinical, pharmaceutical or biomedical procedures
±0.2°C
Accessories