Learn More
Fisherbrand™ Isotemp™ Economy Analog-Control Water Baths
Economical, general-purpose water baths are ideal for industrial, clinical, pharmaceutical and biomedical procedures
Includes:
Sample tray; Thermometer clip; Nonhazardous liquid, partial-immersion (76mm) thermometer (-10° to +110°C); Line cord and grounded NEMA 5-15P plug
Description
- Easy-to-use controls on front panel for full access and safe operation
- Dual thermostats guard against thermal runaway; elements won't burn out even if bath runs dry
- Choice of acrylic or stainless-steel covers
- Hinged stainless-steel or see-through acrylic gable covers have unique fins that keep hands away from hot vapors; stay open at 90° position or lift off to accommodate large glassware
- Corrosion-resistant exterior with acrylic enamel finish; temperature-resistant plastic rim fits snugly to insulate and prevent leaks
- Outside of the bath stays cool to the touch even after hours of operation
- Low-profile design makes access convenient
- Deep-drawn stainless-steel reservoirs are seamless to prevent leaks; rounded corners make cleaning easy
- Shallow-form Model 102S is perfect for heating samples in microcentrifuge tubes and for other molecular biology procedures
- Model 115 has two chambers, each with its own temperature control panel
Order Info
Concentric ring covers and stainless-steel gable covers are available separately.
Service Plan
Service agreement available.
Certifications
CSA approved.
Warranty and Services
One-year warranty on parts, labor and travel.
Specifications
Analog | |
CSA | |
Industrial, clinical, pharmaceutical or biomedical procedures | |
120 | |
1 Year | |
16.88 in. | |
24.38 in. | |
8.88 in. | |
22.25 lb. | |
120 V | |
50/60 Hz |
20 L | |
5°C to 75/100°C | |
Sample tray, thermometer clip, nonhazardous liquid, partial immersion (76mm) thermometer (-10 to 110°C), line cord and grounded NEMA 5-15P plug | |
±0.2°C | |
16.53 x 24.38 x 8.88 in. (42 x 62 x 23 cm) | |
42 cm | |
62 cm | |
23 cm | |
10.1 kg | |
8.3 A |