Learn More
Fisher Scientific™ Isotemp™ Economy Analog-Control Water Baths
Economical, general-purpose water baths are ideal for industrial, clinical, pharmaceutical and biomedical procedures
 View more versions of this product
Catalog No. 15-460-28Q
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
5 to 75/100°C
10.88 in.
50/60Hz
42cm
26 lb.
24.38 in.
8.3A
CSA
Analog
Sample tray, thermometer clip, nonhazardous liquid, partial immersion (76mm) thermometer (-10 to 110°C), line cord and grounded NEMA 5-15P plug
1 Year
120V
28cm
16.88 in.
128
11.8kg
62cm
28L
16.88 x 24.38 x 10.88 in. (42 x 62 x 28cm)
Industrial, clinical, pharmaceutical or biomedical procedures
±0.2°C
Accessories