Fisher Scientific™ Isotemp™ Explosion-Proof Refrigerator

Ideal for storing volatile materials in areas with explosive atmospheres or in hazardous locations

 View more versions of this product

Catalog No. 13-986-153

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Model 20EREEFSA
    Type Explosion Proof Refrigerator
    Control Mechanical Thermostat/Outside, On Top Of Unit, In Blue Housing
    Current 5 Amps/575 Watts
    Casters Adjustable Leveling Feet
    Compressor Bottom Mounted
    Exterior Finish Epoxy Coated Steel
    Interior Finish Epoxy Coated Steel
    Certifications/Compliance cCSAus
    Electrical Requirements 115 V/60 Hz
    Capacity 20.9 Cubic Feet/591.82 Liters
    Defrost Manual Defrost
    Door Count 1(1 compartment)
    Shelves 4 Fixed Shelves, 6 Fixed Door Shelves
    Refrigerant R134a,100% CFC Free
    Door Style Hinged, Opens Left to Right, Non-Reversible
    Net Weight (English) 200 lb.
    Net Weight (Metric) 90.7kg
    Plug Type No Cord Supplied
    Shipping Weight (Metric) 104.3kg
    Color White
    Wattage 575w
    Warranty 13 Months Parts and Labor
    Amperage 5.0A
    Access Security Door lock
    Voltage 115V
    Dimensions (D x W x H) Interior 23.8 x 27.8 x 57.8 in. (60.3 x 70.49 x 146.7cm)
    Capacity (Metric) 566.3L
    Dimensions (L x W x H) Exterior 35.81 x 34 x 73.13 in. (90.95 x 86.4 x 186.1cm)
    Capacity (English) 20 cu. ft.
    Hertz 60Hz
    Insulation Polyurethane Foam
    Shipping Weight (English) 230 lbs.
    Temperature Range (Metric) 1° to 12°C

    Prevent the triggering of an explosion inside or outside the unit. Fisher Scientific™ Isotemp™ Explosion-Proof Refrigerator is suitable for use in Class I, Division I Group C and D hazardous environments. Thermostat and compressor components are isolated within a vapor-proof enclosure, and electrical connections are hardwired to prevent sparking and accidental disconnection.

    Order Info

    Not equipped with line cord and plug; must be hardwired into an appropriately rated conduit box.

    Service Plan

    Service agreement available.

    Certifications

    UL listed meets NFPA 45 and 70

    Warranty

    13 months, parts and labor

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit